Learn More
Invitrogen™ C1QTNF9B Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5103758
Product Code. 16690205
  • £441.00 / 100µL
Description
Antibody detects endogenous levels of total C1QTNF9B.
Probable adipokine. Activates AMPK, AKT, and p44/42 MAPK signaling pathways.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C1QTNF9B
Polyclonal
Unconjugated
C1QTNF9B
C1q and tumor necrosis factor related protein 9B; C1q/TNF-related protein 9B; C1QTNF9B; collagen triple helix repeat-containing; Complement C1q and tumor necrosis factor-related protein 9B; Complement C1q and tumor necrosis factor-related protein 9-like; CTRP9B
Rabbit
Affinity chromatography
RUO
387911
-20°C
Liquid
Western Blot
1 mg/mL
PBS with 50% glycerol and 0.02% sodium azide; pH 7.4
B2RNN3
C1QTNF9B
A synthesized peptide derived from human C1QTNF9B(Accession B2RNN3), corresponding to amino acid residues Y253-L303.
100 μL
Primary
Human, Mouse