missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Novus Biologicals™ C1qTNF5/CTRP5 293T Cell Transient Overexpression Lysate
Shop All Bio Techne Products
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
Specifications
Specifications
| Quantity | 0.1 mL |
| Content And Storage | Store at -80°C. Avoid freeze/thaw cycles. |
| Research Category | Vision |
| Target | C1qTNF5/CTRP5 |
| Synonym | C1q and tumor necrosis factor related protein 5, Complement C1q Tumor Necrosis Factor-Related Protein 5 Precursor Variant 3, CTRP5LORDDKFZp586B0621, LORD 6, Myonectin 3 |
| Gene Symbol | C1QTNF5 |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction