missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
C1qTNF4/CTRP4 Polyclonal antibody specifically detects C1qTNF4/CTRP4 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Specifications
Specifications
| Antigen | C1qTNF4/CTRP4 |
| Applications | Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin) |
| Classification | Polyclonal |
| Conjugate | DyLight 594 |
| Formulation | 50mM Sodium Borate |
| Gene Alias | C1q and tumor necrosis factor related protein 4, complement C1q tumor necrosis factor-related protein 4, complement-c1q tumor necrosis factor-related protein 4, CTRP4ZACRP4 |
| Host Species | Rabbit |
| Immunogen | Antibody was raised against a 16 amino acid synthetic peptide from near the carboxy terminus of human CTRP4. The immunogen is located within amino acids 240 - 290 of CTRP4. Amino Acid Squence: IYDDGASRRREMQSQS |
| Purification Method | Peptide affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?