missing translation for 'onlineSavingsMsg'
Learn More

C1qTNF3/CORS26/CTRP3 Antibody, Janelia Fluor™ 549, Novus Biologicals™

Product Code. 18426577 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
Unit Size:
0.10mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18426577 0.1 mL 0.10mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18426577 Supplier Novus Biologicals Supplier No. NBP176628JF549

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

C1qTNF3/CORS26/CTRP3 Polyclonal antibody specifically detects C1qTNF3/CORS26/CTRP3 in Human, Mouse, Rat samples. It is validated for Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1qTNF3/CORS26/CTRP3
Applications Western Blot, ELISA, Immunohistochemistry, Immunofluorescence, Immunohistochemistry (Paraffin)
Classification Polyclonal
Conjugate Janelia Fluor 549
Formulation 50mM Sodium Borate
Gene Alias C1ATNF3, C1q and tumor necrosis factor related protein 3, cartonectin, collagenous repeat-containing sequence of 26-kDa, complement C1q tumor necrosis factor-related protein 3, complement-c1q tumor necrosis factor-related protein 3, Corcs, Cors, Cors-26, CORS26, CTRP32310005P21Rik, FLJ37576, Secretory protein CORS26
Host Species Rabbit
Immunogen Antibody was raised against a peptide corresponding to 16 amino acids near the center of human C1qTNF3/CORS26/CTRP3. The immunogen is located within amino acids 90-140 of C1qTNF3/CORS26/CTRP3. Amino Acid Sequence: PRGERGQHGPKGEKGY
Purification Method Peptide affinity purified
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 114899
Target Species Human, Mouse, Rat
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.