missing translation for 'onlineSavingsMsg'
Learn More

C1qTNF1/CTRP1 Antibody (395526) [DyLight 550], Novus Biologicals™

Product Code. 30734380 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
0.1 mL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity
30734380 0.1 mL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 30734380 Supplier Novus Biologicals Supplier No. FAB3145L

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C1qTNF1/CTRP1 Monoclonal antibody specifically detects C1qTNF1/CTRP1 in Human samples. It is validated for Western Blot
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1qTNF1/CTRP1
Applications Western Blot
Classification Monoclonal
Clone 395526
Conjugate DyLight 550
Formulation 50mM Sodium Borate
Gene Alias C1q and tumor necrosis factor related protein 1, CTRP1G protein-coupled receptor-interacting protein, FLJ90694, G protein coupled receptor interacting protein, GIPcomplement C1q tumor necrosis factor-related protein 1, ZSIG37
Host Species Mouse
Immunogen Mouse myeloma cell line NS0-derived recombinant human CTRP1/C1qTNF1, Arg26-Pro281, Accession # Q9BXJ1
Purification Method Protein A or G purified from hybridoma culture supernatant
Quantity 0.1 mL
Regulatory Status RUO
Research Discipline Signal Transduction
Primary or Secondary Primary
Gene ID (Entrez) 114897
Target Species Human
Content And Storage Store at 4°C in the dark.
Form Purified
Isotype IgG2b
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.