missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1qR1/CD93 Antibody (R139), mFluor Violet 500 SE, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP1-43378MFV500
This item is not returnable.
View return policy
Description
C1qR1/CD93 Monoclonal antibody specifically detects C1qR1/CD93 in Human samples. It is validated for Flow Cytometry
Specifications
| C1qR1/CD93 | |
| Monoclonal | |
| mFluor Violet 500 SE | |
| 50mM Sodium Borate | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Flow Cytometry | |
| R139 | |
| Flow Cytometry | |
| C1q receptor 1, C1q/MBL/SPA receptor, C1qR(P), C1qRp, CD93 antigenC1qR, CD93 molecule, CDw93C1QR1, Complement component 1 q subcomponent receptor 1, complement component 1, q subcomponent, receptor 1, complement component C1q receptor, dJ737E23.1, ECSM3, Matrix-remodeling-associated protein 4, matrix-remodelling associated 4, MXRA4 | |
| The immunogen for this antibody was CD93. | |
| 0.1 mL | |
| B Cell Development and Differentiation Markers | |
| 22918 | |
| Store at 4C in the dark. | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction