missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1QL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA568157
This item is not returnable.
View return policy
Description
Predicted to react with Mouse samples.
May regulate the number of excitatory synapses that are formed on hippocampus neurons. Has no effect on inhibitory synapses. Plays a role in glucose homeostasis. Via AMPK signaling pathway, stimulates glucose uptake in adipocytes, myotubes and hepatocytes and enhances insulin-stimulated glucose uptake. In a hepatoma cell line, reduces the expression of gluconeogenic enzymes G6PC and PCK1 and hence decreases de novo glucose production. [UniProt].
Specifications
| C1QL3 | |
| Polyclonal | |
| Unconjugated | |
| C1QL3 | |
| 1110065A22Rik; AI661623; C1q and tumor necrosis factor-related protein 13; C1q/TNF-related protein 13; C1ql; C1ql3; C1q-like 3; C1qtnf13; complement C1q like 3; complement C1q-like protein 3; complement component 1, q subcomponent-like 3; CTRP13; Gliacolin; K100 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 227580, 389941 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q5VWW1, Q9ESN4 | |
| C1QL3 | |
| Synthesized peptide derived from human C1QL3. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction