missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QL2 Polyclonal Antibody, Invitrogen™
Rabbit Polyclonal Antibody
Brand: Invitrogen PA563504
This item is not returnable.
View return policy
Description
Immunogen sequence: CRMICDPYTA APGGEPPGAK AQPPGPSTAA LEVMQDLSAN PPPPFIQGPK GD Highest antigen sequence identity to the following orthologs: Mouse - 88%, Rat - 85%.
Specifications
| C1QL2 | |
| Polyclonal | |
| Unconjugated | |
| C1QL2 | |
| CTRP10 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 165257 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q7Z5L3 | |
| C1QL2 | |
| Recombit protein corresponding to Human C1QL2 | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Research Use Only
Spot an opportunity for improvement?Share a Content Correction