missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QL1 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£381.00
Specifications
| Antigen | C1QL1 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
C1QL1 Polyclonal specifically detects C1QL1 in Mouse samples. It is validated for Western Blot.Specifications
| C1QL1 | |
| Polyclonal | |
| Rabbit | |
| O88992 | |
| 10882 | |
| Synthetic peptides corresponding to C1ql1 (complement component 1, q subcomponent-like 1) The peptide sequence was selected from the N terminal of C1ql1. Peptide sequence MLLVLVVLIPVLVSSGGPDGHYEMLGTCRMVCDPYPARGPGAGARSDGGD. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| C1q-related factor, C1QRFCRFC1QTNF14, Complement component 1 Q subcomponent-like 1, complement component 1, q subcomponent-like 1 | |
| C1QL1 | |
| IgG | |
| 28 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title