missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QBP Rabbit anti-Human, Mouse, Rat, CoraLite 488, Clone: 3K20, Proteintech
Rabbit Recombinant Antibody
£135.00 - £375.00
Specifications
| Antigen | C1QBP |
|---|---|
| Clone | 3K20 |
| Applications | Immunofluorescence, Flow Cytometry |
| Conjugate | CoraLite 488 |
| Form | Liquid |
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|---|---|---|---|---|---|---|---|---|---|
| Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
|
17711339
|
Proteintech
CL488-80490-20UL |
20 μL |
£135.00
20µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
|
17689886
|
Proteintech
CL488-80490-100UL |
100 μL |
£375.00
100µL |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
C1QBP antibody apecifically detetects C1QBP antigen in Human, Mouse, Rat Samples. It is validated for Immunofluorescence, Flow Cytometry.Specifications
| C1QBP | |
| Immunofluorescence, Flow Cytometry | |
| Liquid | |
| Human, Mouse, Rat | |
| BC013731 | |
| C1QBP | |
| IgG | |
| Protein A purification |
| 3K20 | |
| CoraLite 488 | |
| Rabbit | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| 708 | |
| C1QBP fusion protein Ag19773 | |
| Primary | |
| Store at -20°C. Avoid exposure to light. |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title