missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QBP Mouse anti-Human, Mouse, CoraLite 488, Clone: 1F9B1, Proteintech
Mouse Monoclonal Antibody
Brand: Proteintech CL488-68084-20UL
This item is not returnable.
View return policy
Description
C1QBP Monoclonal antibody specifically detects C1QBP in Human, Mouse samples. It is validated for IFSpecifications
| C1QBP | |
| Monoclonal | |
| 1000 μg/mL | |
| PBS with 50% Glycerol, 0.05% Proclin300, 0.5% BSA, pH 7.3. | |
| Mouse | |
| 20 μL | |
| 708 | |
| Store at -20°C | |
| Liquid |
| Immunofluorescence | |
| 1F9B1 | |
| CoraLite 488 | |
| BC009600 | |
| Protein A purification | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG2b |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction