missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QBP Monoclonal Antibody (A8F5), Invitrogen™
Mouse Monoclonal Antibody
Brand: Invitrogen MA544925
This item is not returnable.
View return policy
Description
Replication Protein A (RPA) (also known as human single-stranded DNA binding protein, or HSSB) is involved in DNA replication, repair, and recombination. RPA from human cells is a stable heterotrimer of 70kDa, 32-34kDa, and 11-14kDa subunits (RPA70, RPA32, and RPA14 respectively). RPA is required for the SV40 large tumor antigen-catalyzed unwinding of SV40 DNA and stimulates DNA polymerase alpha and delta. RPA34 is phosphorylated at the G1/S boundary of the cell cycle or upon exposure of cells to DNA damage-inducing agents including ionizing and UV radiation.Specifications
C1QBP | |
Monoclonal | |
1.19 mg/mL | |
PBS with 0.1% BSA, 40% glycerol and 0.05% sodium azide; pH 7.4 | |
O35658, O35796, Q07021 | |
C1QBP | |
Recombinant protein within human GC1q R aa 100-282. | |
100 μL | |
Primary | |
Human, Mouse, Rat | |
Liquid |
Flow Cytometry, Immunocytochemistry, Immunohistochemistry (Paraffin), Western Blot | |
A8F5 | |
Unconjugated | |
C1QBP | |
AA407365; AA986492; ASF/SF2-associated protein p32; C1q globular domain-binding protein; C1QBP; complement C1q binding protein; complement component 1 Q subcomponent-binding protein, mitochondrial; complement component 1, q subcomponent binding protein; D11Wsu182e; GC1QBP; gC1qR; gC1Q-R; gC1q-R protein; glycoprotein gC1qBP; HABP1; Hyaluronan-binding protein 1; mitochondrial matrix protein p32; p32; p33; RPA; SF2AP32; SF2p32; splicing factor SF2-associated protein | |
Mouse | |
Protein A | |
RUO | |
12261, 29681, 708 | |
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
IgG2a |