missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1QB Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA551241
This item is not returnable.
View return policy
Description
The antibody detects endogenous levels of total C1QB protein.
C1QB is a major constituent of the human complement subcomponent C1q. C1q associates with C1r and C1s in order to yield the first component of the serum complement system. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
Specifications
| C1QB | |
| Polyclonal | |
| Unconjugated | |
| C1QB | |
| C1QB; C1QTNF5; complement C1q B chain; complement C1q chain B; complement C1q subcomponent subunit B; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component C1q, B chain; complement subcomponent C1q chain B; CTRP5; DKFZP586B0621; LORD | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 12260, 713 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.05% sodium azide; pH 7.4 | |
| P02746, P14106 | |
| C1QB | |
| Fusion protein of human C1QB. | |
| 100 μL | |
| Primary | |
| Human, Mouse | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction