missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1QB, Mouse anti-Human, Clone: C1QB/2966, Novus Biologicals™
Mouse Monoclonal Antibody
Brand: Novus Biologicals NBP2-79788-20UG
This item is not returnable.
View return policy
Description
C1QB Monoclonal specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry-Paraffin, Protein Array.
Specifications
| C1QB | |
| Monoclonal | |
| 0.2 mg/mL | |
| Immunohistochemistry 1-2 ug/ml, Immunohistochemistry-Paraffin 1-2 ug/ml, Protein Array | |
| P02746 | |
| C1QB | |
| Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary) | |
| 20 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry, Immunohistochemistry (Paraffin), Peptide Array | |
| C1QB/2966 | |
| Unconjugated | |
| 10 mM PBS with 0.05% BSA | |
| B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide | |
| Mouse | |
| Protein A or G purified | |
| RUO | |
| 713 | |
| Store at 4°C. | |
| IgG1 κ |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction