missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
C1QB Monoclonal antibody specifically detects C1QB in Human samples. It is validated for Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array
Specifications
Specifications
| Antigen | C1QB |
| Applications | Immunohistochemistry, Immunohistochemistry (Paraffin), Protein Array |
| Classification | Monoclonal |
| Clone | C1QB/2966 |
| Conjugate | APC |
| Formulation | PBS |
| Gene Alias | B chain, complement C1q subcomponent subunit B, complement component 1, q subcomponent, B chain, complement subcomponent C1q chain B, q subcomponent, beta polypeptide |
| Host Species | Mouse |
| Immunogen | Recombinant fragment (around aa 41-188) of human C1QB protein (exact sequence is proprietary) (Uniprot: P02746) |
| Purification Method | Protein A or G purified |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?