Learn More
Invitrogen™ C1q Polyclonal Antibody, FITC
GREENER_CHOICE
Sheep Polyclonal Antibody
Brand:  Invitrogen™ PA184593
Product Code. 11569180
  • £556.00 / 1mL
Description
Sheep anti Human C1q antibody recognizes human C1q, a sub-component of the C1 complement component.
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C1q
Polyclonal
FITC
C1qc
AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B
Sheep
Ion-exchange chromatography
RUO
712, 713, 714
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. Store in the dark.
Liquid
Immunohistochemistry (Frozen)
10 mg/mL
PBS with <0.1% sodium azide; pH 7.2
P02745, P02746, P02747
C1QA, C1QB, C1qc
Human C1q, purified from plasma.
1 mL