missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1q Monoclonal Antibody (7H8)

Rat Monoclonal Antibody
Brand: Invitrogen™ MA140045
This item is not returnable.
View return policy
Description
MA1-40045 detects C1q from mouse samples. MA1-40045 has been successfully used in immunohistochemistry (frozen), ELISA and immunofluorescence applications. The MA1-40045 immunogen is mouse C1q component of the C1 complex.
C1q is a sub-component of complement C1. C1q associates with complement components C1r and C1s, which are required for the progression of the proteolytic complement cascade. Deficiency of C1q has been associated with lupus erythematosus and glomerulonephritis. C1q is composed of 18 polypeptide chains: six A-chains, six B-chains, and six C-chains. Each chain contains a collagen-like region located near the N terminus and a C-terminal globular region. The A-, B-, and C-chains are arranged in the order A-C-B on chromosome 1.
Specifications
| C1q | |
| Monoclonal | |
| 0.1 mg/mL | |
| PBS with 0.1% BSA and 0.02% sodium azide | |
| P14106, P98086, Q02105 | |
| C1QA, C1QB, C1qc | |
| Murine C1q component of the C1 complex. | |
| 100 μg | |
| Primary | |
| Mouse | |
| Antibody | |
| IgG1 |
| ELISA, Immunohistochemistry (Frozen) | |
| 7H8 | |
| Unconjugated | |
| C1QB | |
| AI255395; AI385742; C1q; C1qa; C1qb; C1qc; C1Q-C; C1QG; Ciqc; complement C1q A chain; complement C1q B chain; complement C1q C chain; complement C1q chain A; complement C1q chain B; complement C1q subcomponent subunit A; complement C1q subcomponent subunit A-like protein; Complement C1q subcomponent subunit B; Complement C1q subcomponent subunit C; complement component 1, q subcomponent, A chain; complement component 1, q subcomponent, alpha polypeptide; complement component 1, q subcomponent, B chain; complement component 1, q subcomponent, beta polypeptide; complement component 1, q subcomponent, C chain; complement component 1, q subcomponent, c polypeptide; complement component 1, q subcomponent, gamma polypeptide; complement component C1q, A chain; complement component C1q, B chain; complement subcomponent C1q chain B | |
| Rat | |
| Protein G | |
| RUO | |
| 12259, 12260, 12262 | |
| 4°C | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction