missing translation for 'onlineSavingsMsg'
Learn More

C1q Antibody Pair, Novus Biologicals™

Product Code. 18392068 Shop All Bio Techne Products
Change view
Click to view available options
Quantity:
1 Set
Unit Size:
1 set
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
18392068 1 Set 1 set
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 18392068 Supplier Novus Biologicals Supplier No. H00000712AP21

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Antibody Pair

C1q Polyclonal antibody specifically detects C1q in Human samples. It is validated for ELISA
TRUSTED_SUSTAINABILITY

Specifications

Antigen C1q
Applications Sandwich ELISA
Classification Polyclonal
Conjugate Unconjugated
Dilution Sandwich ELISA 3 to 100 ng/mL
Formulation 1x PBS, pH 7.4
Gene Alias A chain, complement component 1, q subcomponent, A chain, complement component 1, q subcomponent, alpha polypeptide
Quantity 1 Set
Regulatory Status RUO
Research Discipline Immunology, Innate Immunity
Primary or Secondary Primary
Gene ID (Entrez) 712
Target Species Human
Content And Storage Aliquot and store at -20°C or -80°C. Avoid freeze-thaw cycles.
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.