missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf85 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA556060
This item is not returnable.
View return policy
Description
Immunogen sequence: VSLEVIPNWL GPLQNLLHIR AVGTNSTLHY VWSSLGPLAV VMVATNTPHS TLSVNWSLLL SPEPDGGLMV LPKDSIQFSS ALVFTRLLEF DSTNVSDTAA KPLGRPYPPY SLADFSWNNI TDSLDPATLS ATFQGHPMND PTRT Highest antigen sequence identity to the following orthologs: Mouse - 77%, Rat - 76%.
Specifications
| C1orf85 | |
| Polyclonal | |
| Unconjugated | |
| GLMP | |
| 0610031j06rik; AB027141; C1orf85; GLMP; Glycosylated lysosomal membrane protein; kidney lysosomal membrane protein; kidney predominant protein NCU-G1; lysosomal protein NCU-G1; NCU-G1; PSEC0030; RGD1303130; UNQ2553/PRO6182 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 112770 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q8WWB7 | |
| GLMP | |
| Recombinant protein corresponding to Human C1orf85. Recombinant protein control fragment (Product #RP-90590). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction