Learn More
Invitrogen™ C1orf85 Polyclonal Antibody
GREENER_CHOICE
Goat Polyclonal Antibody
Brand:  Invitrogen™ PA5142985
Product Code. 17875662
  • £414.00 / 100µL
Description
Additional Information: The immunizing peptide represents the luminal part of the protein. This antibody is tested in Peptide ELISA: antibody detection limit dilution 1:32,000.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C1orf85
Polyclonal
Unconjugated
GLMP
0610031j06rik; AB027141; C1orf85; GLMP; Glycosylated lysosomal membrane protein; kidney lysosomal membrane protein; kidney predominant protein NCU-G1; lysosomal protein NCU-G1; NCU-G1; PSEC0030; RGD1303130; UNQ2553/PRO6182
Goat
Ammonium sulfate precipitation
RUO
295231, 56700
-20°C, Avoid Freeze/Thaw Cycles
Liquid
Western Blot
0.5 mg/mL
TBS with 0.5% BSA and 0.02% sodium azide; pH 7.3
Q68FV6, Q9JHJ3
GLMP
Peptide with sequence PDCPSVNERNSIDD (aa 243-256).
100 μg
Primary
Mouse, Rat