missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C1orf50 Polyclonal Antibody
GREENER_CHOICE

Rabbit Polyclonal Antibody

Brand:  Invitrogen™ PA556411

Product Code. 15754321

  • £467.00 / 100µL

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

Immunogen sequence: HHVACNIVKK PGNIYYLYKR ESGQQYFSII SPKEWGTSCP HDFLGAYKLQ HDLSWTPYED IEKQDAKISM MDTLLSQSVA LPPCTEPNFQ GLTH Highest antigen sequence identity to the following orthologs: Mouse - 86%, Rat - 86%.

C1orf50 is a protein coding gene.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C1orf50
Polyclonal
Unconjugated
C1orf50
C15H1orf50; C1orf50; chromosome 1 open reading frame 50; chromosome 15 C1orf50 homolog; uncharacterized protein C1orf50; uncharacterized protein C1orf50 homolog
Rabbit
Antigen affinity chromatography
RUO
79078
Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Liquid
Immunohistochemistry (Paraffin), Western Blot
0.1 mg/mL
PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Q9BV19
C1orf50
Recombinant protein corresponding to Human C1orf50. Recombinant protein control fragment (Product #RP-94442).
100 μL
Primary
Human
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.