missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf43 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA544182
This item is not returnable.
View return policy
Description
Peptide sequence: YQEALSELAT AVKARIGSSQ RHHQSAAKDL TQSPEVSPTT IQVTYLPSSQ Sequence homology: Cow: 93%; Dog: 93%; Guinea Pig: 93%; Horse: 93%; Human: 100%; Mouse: 93%; Rabbit: 93%; Rat: 93%.
General regulator of phagocytosis. Required to uptake Gram negative bacterium by macrophages. [UniProt].
Specifications
| C1orf43 | |
| Polyclonal | |
| Unconjugated | |
| C1orf43 | |
| C1orf43; chromosome 1 open reading frame 43; HCV NS5A-transactivated protein 4; Hepatitis C virus NS5A-transactivated protein 4; HSPC012; NICE3; NICE-3; NS5ATP4; Protein NICE-3; S863-3; Uncharacterized protein C1orf43 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 25912 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| Western Blot | |
| 0.5 mg/mL | |
| PBS with 2% sucrose and 0.09% sodium azide | |
| Q9BWL3 | |
| C1orf43 | |
| Synthetic peptide directed towards the middle region of human C1orf43. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction