missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf35 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5101249
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total MMTAG2.
MMTAG2 gene ontology (GO) annotations include neutrophil degranulation.
Specifications
| C1orf35 | |
| Polyclonal | |
| Unconjugated | |
| C1orf35 | |
| 2310033P09Rik; C1orf35; chromosome 1 open reading frame 35; hMMTAG2; MMTAG2; multiple myeloma transforming 2; multiple myeloma tumor-associated protein 2; Multiple myeloma tumor-associated protein 2 homolog; RGD1304587; RIKEN cDNA 2310033P09 gene; similar to RIKEN cDNA 2310033P09 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 67862, 79169 | |
| -20°C | |
| Liquid |
| Western Blot, Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q99LX5, Q9BU76 | |
| 2310033P09Rik, C1orf35 | |
| A synthesized peptide derived from human MMTAG2, corresponding to a region within C-terminal amino acids. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction