missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf27 Rabbit anti-Human, Mouse, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
£133.90 - £349.17
Specifications
| Antigen | C1orf27 |
|---|---|
| Concentration | 0.35 mg/mL |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
Description
May play a role in the trafficking of a subset of G-protein coupled receptors.Specifications
| C1orf27 | |
| Western Blot | |
| Unconjugated | |
| Rabbit | |
| Human, Mouse | |
| Q4PJX1, Q5SWX8 | |
| 226499, 54953 | |
| C1orf27 Fusion Protein Ag18802 | |
| Primary | |
| -20°C | |
| ODR4 |
| 0.35 mg/mL | |
| Polyclonal | |
| Liquid | |
| RUO | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| C1orf27, hODR 4, LAG1 interacting protein, odr 4, ODR4, Protein odr 4 homolog, TTG1, TTG1A | |
| ODR4 | |
| IgG | |
| Antigen Affinity Chromatography | |
| Antibody |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title