missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf226 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA562576
This item is not returnable.
View return policy
Description
Immunogen sequence: DFLRRKEPSS LGSVGVTEIN KTAGAQLASG TDAAPEAWLE DERSVLQETF PRLDPPPPIT RKRTPRALKT TQD Highest antigen sequence identity to the following orthologs: Mouse - 84%, Rat - 81%.
C1orf226 (Chromosome 1 Open Reading Frame 226) is a Protein Coding gene. Diseases associated with C1orf226 include Phosphoglycerate Dehydrogenase Deficiency. An important paralog of this gene is DAB2.
Specifications
| C1orf226 | |
| Polyclonal | |
| Unconjugated | |
| C1orf226 | |
| C1orf226; chromosome 1 open reading frame 226; uncharacterized protein C1orf226 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 400793 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 0.2 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| A1L170 | |
| C1orf226 | |
| Recombinant protein corresponding to Human C1orf226. Recombinant protein control fragment (Product #RP-94567). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction