missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf167 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP3-06367-100ul
This item is not returnable.
View return policy
Description
C1orf167 Polyclonal antibody specifically detects C1orf167 in Human samples. It is validated for Immunohistochemistry (Paraffin)
Specifications
| C1orf167 | |
| Polyclonal | |
| Immunohistochemistry-Paraffin 1:200-1:1000 | |
| chromosome 1 open reading frame 167, DKFZp434E1410, DKFZp434F1313, hypothetical protein LOC284498 | |
| Produced in rabbits immunized with E. coli-derived Human C1orf167 fragment. | |
| 100 μg | |
| Primary | |
| Human | |
| Purified |
| Immunohistochemistry (Paraffin) | |
| Unconjugated | |
| PBS, pH7.0 | |
| Rabbit | |
| Antigen and protein A Affinity-purified | |
| RUO | |
| 284498 | |
| Store at 4°C short term. Aliquot and store at -20°C long term. Avoid freeze-thaw cycles. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction