missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf146 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
Brand: Novus Biologicals NBP2-42820
This item is not returnable.
View return policy
Description
Specifically detects C1orf146 in Human samples, and it is validated for Western Blotting
C1orf146 Polyclonal specifically detects C1orf146 in Human samples. It is validated for Western Blot.
Specifications
| C1orf146 | |
| Polyclonal | |
| Western Blot 1:500-1:3000 | |
| SCRE, SPO16 | |
| Recombinant protein encompassing a sequence within the center region of human C1orf146. The exact sequence is proprietary. | |
| 0.1 mL | |
| Primary | |
| Human | |
| IgG |
| Western Blot | |
| Unconjugated | |
| 0.1M Tris (pH 7.0), 0.1M Glycine and 20% Glycerol with 0.01% Thimerosal | |
| Rabbit | |
| Affinity Purified | |
| RUO | |
| 388649 | |
| Aliquot and store at -20C or -80C. Avoid freeze-thaw cycles. |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
For Research Use Only
Spot an opportunity for improvement?Share a Content Correction