missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1orf122 Antibody, Novus Biologicals™
Rabbit Polyclonal Antibody
£362.00
Specifications
| Antigen | C1orf122 |
|---|---|
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Host Species | Rabbit |
Description
C1orf122 Polyclonal specifically detects C1orf122 in Human samples. It is validated for Western Blot.Specifications
| C1orf122 | |
| Polyclonal | |
| Rabbit | |
| Human | |
| 127687 | |
| Synthetic peptides corresponding to C1orf122 () The peptide sequence was selected from the n terminal of C1orf122)(50ug). Peptide sequence GAGARREATPTAAPGGARQAAAGRGGAAAAAAPGHHRSLRGDVTAAGPPA. | |
| Primary |
| Western Blot | |
| Unconjugated | |
| RUO | |
| ALAESM, chromosome 1 open reading frame 122 | |
| C1ORF122 | |
| IgG | |
| 16 kDa |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title