missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1orf111 Monoclonal Antibody (OTI2A9)

Mouse Monoclonal Antibody
Brand: Invitrogen™ MA527389
This item is not returnable.
View return policy
Description
C1orf111 Monoclonal Antibody for Western Blot, IHC (P)
Plays a role in spermiogenesis and fertilization. [UniProt].
Specifications
| C1orf111 | |
| Monoclonal | |
| 1 mg/mL | |
| PBS with 1% BSA, 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q5T0L3 | |
| SPATA46 | |
| Full length human recombinant protein of C1ORF111 produced in E.coli. | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG1 |
| Immunohistochemistry (Paraffin), Western Blot | |
| OTI2A9 | |
| Unconjugated | |
| SPATA46 | |
| 1700015E13Rik; C1orf111; chromosome 1 open reading frame 111; HSD20; RGD1565283; RIKEN cDNA 1700015E13 gene; RP11-565P22.3; similar to novel protein; SPATA46; Spermatogenesis-associated protein 46; uncharacterized protein C1orf111; uncharacterized protein C1orf111 homolog | |
| Mouse | |
| Affinity chromatography | |
| RUO | |
| 284680 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction