Learn More
Invitrogen™ C1orf101 Polyclonal Antibody
GREENER_CHOICE
Rabbit Polyclonal Antibody
Brand:  Invitrogen™ PA5112984
Product Code. 17150844
  • £405.00 / 100µg
Description
C1orf101 Polyclonal Antibody for Western Blot, IHC (P), ICC/IF, ELISA
Auxiliary component of the CatSper complex, a complex involved in sperm cell hyperactivation. Sperm cell hyperactivation is needed for sperm motility which is essential late in the preparation of sperm for fertilization. [UniProt].
TRUSTED_SUSTAINABILITY
Specifications
Specifications
C1orf101
Polyclonal
Unconjugated
CATSPERE
C1orf101; Cation channel sperm-associated protein subunit epsilon; CATSPERE; CatSper-epsilon; chromosome 1 open reading frame 101; uncharacterized protein C1orf101
Rabbit
Protein G
RUO
257044
-20°C or -80°C if preferred
Liquid
ELISA, Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry
3.97 mg/mL
PBS with 50% glycerol and 0.03% ProClin 300; pH 7.4
Q5SY80
CATSPERE
Recombinant Human Cation channel sperm-associated protein subunit epsilon protein (601-832AA).
100 μg
Primary
Human, Mouse