missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C1GALT1C1 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5100451
This item is not returnable.
View return policy
Description
Antibody detects endogenous levels of total COSMC.
This gene encodes a type II transmembrane protein that is similar to the core 1 beta1,3-galactosyltransferase 1, which catalyzes the synthesis of the core-1 structure, also known as Thomsen-Friedenreich antigen, on O-linked glycans. This gene product lacks the galactosyltransferase activity itself, but instead acts as a molecular chaperone required for the folding, stability and full activity of the core 1 beta1,3-galactosyltransferase 1. Mutations in this gene have been associated with Tn syndrome. Alternatively spliced transcript variants encoding the same protein have been identified.
Specifications
| C1GALT1C1 | |
| Polyclonal | |
| Unconjugated | |
| C1GALT1C1 | |
| 1500002I11Rik; AB030184; beta 1,3-galactosyltransferase 2; beta1,3-galactosyltransferase 2; C1GALT1 specific chaperone 1; C1galt1c1; C1GALT1-specific chaperone 1; C1GALT2; C1Gal-T2; C38H2-L1; C38H2-like protein 1; C81205; core 1 beta1,3-galactosyltransferase 2; core 1 beta3-galactosyltransferase-specific molecular chaperone; core 1 beta3-Gal-T2; core 1 UDP-galactose:N-acetylgalactosamine-alpha-R beta 1,3-galactosyltransferase 2; COSMC; HSPC067; mC1Gal-T2; MST143; MSTP143; RGD1311230; TNPS; UNQ273/PRO310 | |
| Rabbit | |
| Affinity chromatography | |
| RUO | |
| 29071, 302499, 59048 | |
| -20°C | |
| Liquid |
| Western Blot | |
| 1 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.4 | |
| Q499P3, Q96EU7, Q9JMG2 | |
| C1GALT1C1 | |
| A synthesized peptide derived from human C1GALT1C1(Accession Q96EU7), corresponding to amino acid residues H93-A143. | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction