missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C1D Antibody (004), HRP, Novus Biologicals™
Rabbit Monoclonal Antibody
Brand: Novus Biologicals NBP2-90103H
This item is not returnable.
View return policy
Description
C1D Monoclonal specifically detects C1D in Human samples. It is validated for Western Blot.
Specifications
| C1D | |
| Monoclonal | |
| HRP | |
| PBS | |
| Rabbit | |
| Protein A purified | |
| RUO | |
| Primary | |
| Human | |
| Purified |
| Western Blot | |
| 4 | |
| Western Blot | |
| C1D DNA-binding protein, C1D nuclear receptor corepressor, C1D nuclear receptor co-repressor, hC1D, MGC12261, MGC14659, nuclear DNA-binding protein, nuclear nucleic acid-binding protein C1D, small unique nuclear receptor corepressor, small unique nuclear receptor co-repressor, SUN-CoR, SUNCORLRP1 | |
| This antibody was obtained from a rabbit immunized with purified, recombinant Human C1D (Q13901; Met1-Ser141). | |
| 0.1 mL | |
| DNA replication Transcription Translation and Splicing | |
| 10438 | |
| Store at 4°C in the dark. | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction