missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C19orf39 Polyclonal Antibody

C19orf39 Polyclonal Antibody
Brand: Thermo Scientific PA562669
This item is not returnable.
View return policy
Description
C19orf39 Polyclonal Antibody for IHC
Specifications
| C19orf39 | |
| Polyclonal | |
| Unconjugated | |
| SWSAP1 | |
| 2310047B19Rik; ATPase SWSAP1; C19orf39; SWIM type zinc finger 7 associated protein 1; SWIM-type zinc finger 7 associated protein 1; SWIM-type zinc finger 7-associated protein 1; SWS1AP1; SWS1-associated protein 1; Swsap1; zinc finger, SWIM-type containing 7 associated protein 1; ZSWIM7AP1; ZSWIM7-associated protein 1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 126074 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6NVH7 | |
| SWSAP1 | |
| Recombinant protein corresponding to Human C19orf39. Recombinant protein control fragment (Product #RP-99179). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction