missing translation for 'onlineSavingsMsg'
Learn More

PeproTech C19orf10 Polyclonal Antibody, Biotin, PeproTech®, Invitrogen™

Rabbit Polyclonal Antibody

Brand:  PeproTech 500-P259BT-500UG

 View more versions of this product

Product Code. 30054433

  • £1466.00 / 500µg

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Description

Description

AA Sequence of recombinant protein: MVSEPTTVPF DVRPGGVVHS FSQDVGPGNK FTCTFTYASQ GGTNEQWQMS LGTSEDSQHF TCTIWRPQGK SYLYFTQFKA ELRGAEIEYA MAYSKAAFER ESDVPLKSEE FEVTKTAVSH RPGAFKAELS KLVIVAKAAR SEL. Preparation: Produced from sera of rabbits immunized with highly pure Recombinant Murine SF-20. Anti-Murine SF-20-specific antibody was purified by affinity chromatography and then biotinylated. Sandwich ELISA: To detect mSF-20 by sandwich ELISA (using 100 μL/well antibody solution) a concentration of 0.25-1.0 μg/mL of this antibody is required. This biotinylated polyclonal antibody, in conjunction with PeproTech Polyclonal Anti-Murine SF-20 (500-P259) as a capture antibody, allows the detection of at least 0.2-0.4 ng/well of Recombinant mSF-20. Western Blot: To detect mSF-20 by Western Blot analysis this antibody can be used at a concentration of 0.1-0.2 μg/mL. Used in conjunction with compatible secondary reagents the detection limit for Recombinant mSF-20 is 1.5-3.0 ng/lane, under either reducing or non-reducing conditions. 500-P259BT-1MG will be provided as 2 x 500 μg

The interleukins are a broad family of well characterized cytokines, primarily of hematopoietic cell origin. Cytokines are small, soluble proteins with pleiotropic effects on a variety of cell types. Cytokines have a regulatory function over the immune system and mediate aspects of inflammatory response. IL-25 (also called SF20/IL-25) is a secreted bone marrow stroma-derived growth factor cytokine that is derived from Th2 T-cells. IL-25 belongs to the IL-17 cytokine family and is capable of amplifying allergic type inflammatory responses by its actions on other cell types. IL-25 binds to mouse thymic shared antigen-1 and supports lymphoid cell proliferation. Infusion of mice with IL-25 induces IL-4, IL-5, and IL-13 gene expression. IL-25 induced mice also develop epithelial cell hyperplasia, increased mucus secretion, and airway hyperreactivity, suggesting that IL-25 may be an important mediator of allergic disease via production of IL-4, IL-5, IL-13 and eotaxin.
TRUSTED_SUSTAINABILITY
Specifications

Specifications

C19orf10
Polyclonal
Biotin
MYDGF
C19orf10; CS010; D17Wsu104e; EUROIMAGE1875335; IL 25; IL 27; Il25; IL27; IL27w; interleukin 25; interleukin 27 working designation; Interleukin25; interleukin-25; LOC501282; Ly6elg; Mydgf; myeloid derived growth factor; myeloid-derived growth factor; R33729 1; R33729_1; R337291; SF20; similar to lymphocyte antigen 6 complex, locus E ligand; stromal cell-derived growth factor SF20; UPF0556 protein C19orf10; UPF0556 protein C19orf10 homolog
Rabbit
Antigen affinity chromatography
RUO
28106
-20°C
Lyophilized
ELISA, Western Blot
0.1-1.0 mg/mL
PBS with no preservative
Q9CPT4
MYDGF
E.coli-derived, 15.8 kDa Recombinant Murine SF-20.
500 μg
Primary
Mouse
Antibody
IgG
Product Suggestions

Product Suggestions

Videos
SDS
Documents

Documents

Certificates
Promotions

Promotions

Product Content Correction

Your input is important to us. Please complete this form to provide feedback related to the content on this product.

Product Title

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.

Thank You! Your feedback has been submitted. Fisher Scientific is always working to improve our content for you. We appreciate your feedback.