missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C18orf8 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20111-1-AP-150UL
This item is not returnable.
View return policy
Description
Colon cancer-associated protein Mic1.Specifications
| C18orf8 | |
| Polyclonal | |
| Unconjugated | |
| C18orf8 | |
| C18orf8, HsT2591, Mic 1, MIC1 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 291784, 29919, 76482 | |
| -20°C | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.19 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q8VC42, Q96DM3 | |
| C18orf8, Rmc1 | |
| C18orf8 Fusion Protein Ag13904 | |
| 150 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction