missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
Immunogen sequence: STSSSDDDEE VSGSSKTITA EIPGHLDPGF LASDKTSAGN APLNEEINIA For short term use, store at 2-8°C up to 1 week. For long term storage, store at -20°C in small aliquots to prevent freeze-thaw cycles. Predicted homology: Cow: 100%; Dog: 100%; Horse: 100%; Human: 100%; Mouse: 100%; Pig: 100%; Rabbit: 100%; Rat: 100%.
The exact function of C18orf25 remains unknown. There are two named isoforms.
Specifications
Specifications
| Antigen | C18orf25 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Concentration | 0.5 mg/mL |
| Conjugate | Unconjugated |
| Formulation | PBS with 2% sucrose and 0.09% sodium azide |
| Gene | C18orf25 |
| Gene Accession No. | Q96B23 |
| Gene Alias | 8030462N17Rik; Akd2; Ark2N; arkadia 2; ARKadia-like 1; ARKadia-like protein 1; ARKL1; C18orf25; chromosome 18 open reading frame 25; cr025; RIKEN cDNA 8030462N17 gene; RNF111L1; Uncharacterized protein C18orf25; uncharacterized protein C18orf25 homolog |
| Gene Symbols | C18orf25 |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?
