missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C17orf99 Polyclonal Antibody
GREENER_CHOICE

Product Code. 15794131 Shop All Thermo Scientific Products
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
15794131 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 15794131 Supplier Invitrogen™ Supplier No. PA556224

Please to purchase this item. Need a web account? Register with us today!

This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products

This item is not returnable. View return policy

Rabbit Polyclonal Antibody

Immunogen sequence: FSFLPSQTSD WFWCQAANNA NVQHSALTVV PPGGDQKMED WQGPLESPIL ALPLYRSTRR LSEEEFGGFR IGNGEVRGRK AAAM Highest antigen sequence identity to the following orthologs: Mouse - 26%, Rat - 23%.

Probable B cell-associated cytokine that plays a role in the regulation of humoral immune responses. Involved in lymphocyte B cell development and immunoglobulin/IgA production.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C17orf99
Applications Immunohistochemistry (Paraffin)
Classification Polyclonal
Concentration 0.5 mg/mL
Conjugate Unconjugated
Formulation PBS with 40% glycerol and 0.02% sodium azide; pH 7.2
Gene C17orf99
Gene Accession No. Q6UX52
Gene Alias C17orf99; chromosome 17 open reading frame 99; GLPG464; IL40; IL-40; Interleukin-40; Protein IL-40; uncharacterized protein C17orf99; UNQ464; UNQ464/PRO809
Gene Symbols C17orf99
Host Species Rabbit
Immunogen Recombinant protein corresponding to Human C17orf99. Recombinant protein control fragment (Product #RP-93530).
Purification Method Antigen affinity chromatography
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 100141515
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.