missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C17orf80 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5116363
This item is not returnable.
View return policy
Description
C17orf80 Polyclonal Antibody for Western Blot
Specifications
| C17orf80 | |
| Polyclonal | |
| PBS with 0.09% sodium azide; pH 7.4 | |
| Q9BSJ5 | |
| C17orf80 | |
| KLH conjugated synthetic peptide between 264-696 amino acids from the Central region of human C17orf80. | |
| 400 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
| Western Blot | |
| Unconjugated | |
| C17orf80 | |
| C17orf80; Cell migration-inducing gene 3 protein; chromosome 17 open reading frame 80; HLC8; HLC-8; Human lung cancer oncogene 8 protein; lung cancer-related protein 8; MIG3; SPEP1; sperm-expressed protein 1; uncharacterized protein C17orf80 | |
| Rabbit | |
| Antigen affinity chromatography, Protein A | |
| RUO | |
| 55028 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction