Learn More
Invitrogen™ C17orf70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA5121212
Description
Positive test controls include: SH-SY5Y, A-431, HeLa, Mouse brain, Mouse liver, Rat brain. The target is usually found in the following locations: Nucleus. Immunogen sequence: CLRFPGLAPP HTRAPSPLGP TRDPVATFLE TCREPGSQPA GPASLRAEYL PPSVASIKVS AELLRAALKD GHSGVPLCCA TLQWLLAENA AVDVVRARAL SSIQGVAPDG ANVHLIVREV AMTDLCPAGP IQAVEIQVES SSLADICRAH HAVVGRMQTM VTEQATQGSS APDLRVQYLR QIHANHETLL REVQTLRDRL CTEDEASSCA TAQRLLQVYR QLRHPSLILL.
FAAP100 is a component of the Fanconi anemia (FA; MIM 277650) core complex and is required for core complex stability and FANCD2 (see MIM 227646) monoubiquitination (Ling et al., 2007 [PubMed 17396147]).
Specifications
| C17orf70 | |
| Polyclonal | |
| Unconjugated | |
| FAAP100 | |
| 2310003H01Rik; C17orf70; FAAP100; Fanconi anemia associated protein 100 kDa subunit; Fanconi anemia core complex 100 kDa subunit; Fanconi anemia core complex associated protein 100; Fanconi anemia core complex-associated protein 100; Fanconi anemia-associated protein of 100 kDa; Fanconi anemia-associated protein, 100kDa; RGD1306926; RIKEN cDNA 2310003H01 gene; similar to hypothetical protein FLJ22175 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 303742, 71885, 80233 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot | |
| 0.81 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| A2ACJ2, Q0VG06 | |
| FAAP100 | |
| Recombinant fusion protein containing a sequence corresponding to amino acids 652-881 of human C17orf70 (NP_079437.5). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.