missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C17orf70 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA555334
This item is not returnable.
View return policy
Description
Immunogen sequence: VLCSVSPSGS RVPHDLLGGS GGFTLEDALF GLLFGADATL LQSPVVLCGL PDGQLCCVIL KALVTSRSAP GDPNALVKIL HHLEEPVIFI GALKTEPQAA EAAENFLPDE DVHCDC Highest antigen sequence identity to the following orthologs: Mouse - 72%, Rat - 71%.
FAAP100 is a component of the Fanconi anemia (FA; MIM 277650) core complex and is required for core complex stability and FANCD2 (see MIM 227646) monoubiquitination (Ling et al., 2007 [PubMed 17396147]).
Specifications
| C17orf70 | |
| Polyclonal | |
| Unconjugated | |
| FAAP100 | |
| 2310003H01Rik; C17orf70; FAAP100; Fanconi anemia associated protein 100 kDa subunit; Fanconi anemia core complex 100 kDa subunit; Fanconi anemia core complex associated protein 100; Fanconi anemia core complex-associated protein 100; Fanconi anemia-associated protein of 100 kDa; Fanconi anemia-associated protein, 100kDa; RGD1306926; RIKEN cDNA 2310003H01 gene; similar to hypothetical protein FLJ22175 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 80233 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q0VG06 | |
| FAAP100 | |
| Recombinant protein corresponding to Human C17orf70. Recombinant protein control fragment (Product #RP-93510). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction