missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C17orf64 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA560703
This item is not returnable.
View return policy
Description
Immunogen sequence: LHSNISGMKE RLSNMQTPGQ GSPLPGQPRS QDHVKKDSLR ELSQKPKLKR KRIKEAPETP ETE Highest antigen sequence identity to the following orthologs: Mouse - 60%, Rat - 62%.
Specifications
| C17orf64 | |
| Polyclonal | |
| Unconjugated | |
| C17orf64 | |
| C17orf64; chromosome 17 open reading frame 64; uncharacterized protein C17orf64 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 124773 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q86WR6 | |
| C17orf64 | |
| Recombinant protein corresponding to Human C17orf64. Recombinant protein control fragment (Product #RP-93291). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction