missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C16orf89 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA553071
This item is not returnable.
View return policy
Description
Immunogen sequence: KSVREKWAQE PLLQPLSLRV GMLGEKLEAA IQRSLHYLKL SDPKYLREFQ LTLQPGFWKL PHAWIHTDAS LVYPTFGPQD SFSEERSDVC LVQLLGTGTD SSEPCGLSDL CRSLMTKPGC SGYCLSHQLL FFLWA Highest antigen sequence identity to the following orthologs: Mouse - 70%, Rat - 73%.
This gene is expressed predominantly in the thyroid. Based on expression patterns similar to thyroid transcription factors and proteins, this gene may function in the development and function of the thyroid. Multiple transcript variants encoding different isoforms have been found for this gene. [provided by RefSeq, Oct 2011].
Specifications
| C16orf89 | |
| Polyclonal | |
| Unconjugated | |
| C16orf89 | |
| C16orf89; chromosome 16 open reading frame 89; MGC45438; UNQ904/PRO1925; UPF0764 protein C16orf89 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 146556 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin) | |
| 0.05 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6UX73 | |
| C16orf89 | |
| Recombinant protein corresponding to Human C16orf89. Recombinant protein control fragment (Product #RP-89929). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction