Learn More
Invitrogen™ GTL3 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA596577
Description
Immunogen sequence: LDDKNVRRRF RASNYQSTTR VKPFICTMPM RLDDGWNQIQ FNLLDFTRRA YGTNYIETLR VQIHANCRIR RVYFSDRLYS EDELPAEFKL YLPVQNKAKQ Positive Samples: SW620, NCI-H460, Jurkat, SKOV3, HepG2, Mouse lung, Mouse liver, Rat lung; Cellular Location: Cell projection, Cytoplasm, Nucleus, centriole, centrosome, cilium, cilium basal body, cytoskeleton, microtubule organizing center.
CFAP20 (Cilia And Flagella Associated Protein 20) is a Protein Coding gene. Diseases associated with CFAP20 includeFamilial Atrial FibrillationandMaturity-Onset Diabetes Of The Young, Type 1.
Specifications
| GTL3 | |
| Polyclonal | |
| Unconjugated | |
| CFAP20 | |
| 2600014O15Rik; AL024127; Basal body up-regulated protein 22; BUG22; C16orf80; C18H16orf80; C2H16orf80; C3H16orf80; C5H16orf80; CFAP20; cilia and flagella associated protein 20; cilia- and flagella-associated protein 20; EVORF; flagellar associated protein 20 homolog; fSAP23; functional spliceosome-associated protein 23; gene trap locus 3; gene trap locus 3 protein; Gene trap locus 3 protein homolog; GTL3; likely ortholog of mouse gene trap locus 3; similar to transcription factor IIB; T10-2A2; transcription factor 2B; transcription factor IIB; UPF0468 protein C16orf80; UPF0468 protein C16orf80 homolog; zgc:73380 | |
| Rabbit | |
| Affinity Chromatography | |
| RUO | |
| 14894, 29105, 307642 | |
| -20°C, Avoid Freeze/Thaw Cycles | |
| Liquid |
| ELISA, Western Blot, Immunocytochemistry | |
| 1.52 mg/mL | |
| PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
| Q499T7, Q8BTU1, Q9Y6A4 | |
| CFAP20 | |
| A synthetic peptide corresponding to a sequence within amino acids 94-193 of human C16orf80 (NP_0373741). | |
| 100 μL | |
| Primary | |
| Human, Mouse, Rat | |
| Antibody | |
| IgG |
Your input is important to us. Please complete this form to provide feedback related to the content on this product.