missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Description
C16orf58 Polyclonal specifically detects C16orf58 in Human samples. It is validated for Western Blot.
Specifications
Specifications
| Antigen | C16orf58 |
| Applications | Western Blot |
| Classification | Polyclonal |
| Conjugate | Unconjugated |
| Formulation | PBS, 2% Sucrose |
| Gene Alias | chromosome 16 open reading frame 58, FLJ13868, hypothetical protein LOC64755 |
| Host Species | Rabbit |
| Immunogen | The immunogen is a synthetic peptide directed towards the middle region of human C16orf58 |
| Purification Method | Immunogen affinity purified |
| Quantity | 0.1 mL |
| Show More |
Product Title
By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.
Spot an opportunity for improvement?