missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf58 Polyclonal Antibody

C16orf58 Polyclonal Antibody
Brand: Thermo Scientific PA555071
This item is not returnable.
View return policy
Description
C16orf58 Polyclonal Antibody for Western Blot, ICC/IF, IHC
Specifications
| C16orf58 | |
| Polyclonal | |
| Unconjugated | |
| C16orf58 | |
| C16orf58; chromosome 16 open reading frame 58; RUS; RUS1 family protein C16orf58; UPF0420 protein C16orf58 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 64755 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q96GQ5 | |
| C16orf58 | |
| Recombinant protein corresponding to Human C16orf58. Recombinant protein control fragment (Product #RP-90278). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction