missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C16orf57 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559732
This item is not returnable.
View return policy
Description
Immunogen sequence: LVSEVDRVME EFNLTTFYQD PSFHLSLAWC VGDARLQLEG QCLQELQAIV DGFEDAEVLL RVHTEQVRCK SGNKFFSM Highest antigen sequence identity to the following orthologs: Mouse - 85%, Rat - 86%.
C16orf57 is a phosphodiesterase responsible for the U6 snRNA 3' end processing. This protein acts as an exoribonuclease (RNase) responsible for trimming the poly(U) tract of the last nucleotides in the pre-U6 snRNA molecule, leading to the formation of mature U6 snRNA 3' end-terminated with a 2',3'-cyclic phosphate.
Specifications
| C16orf57 | |
| Polyclonal | |
| Unconjugated | |
| USB1 | |
| AA960436; C16orf57; FLJ13154; HAMAP-Rule:MF_03040}; hUsb1; HVSL motif containing 1; HVSL1; Mpn1; mutated in poikiloderma with neutropenia protein 1; PN; putative U6 snRNA phosphodiesterase; RGD1305215; U six biogenesis 1; U6 snRNA biogenesis 1; U6 snRNA biogenesis phosphodiesterase 1; U6 snRNA phosphodiesterase; U6 snRNA phosphodiesterase {ECO:0000255; UPF0406 protein C16orf57; UPF0406 protein C16orf57 homolog; USB1 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 79650 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot | |
| 0.3 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q9BQ65 | |
| USB1 | |
| Recombinant protein corresponding to Human USB1. Recombinant protein control fragment (Product #RP-98621). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction