missing translation for 'onlineSavingsMsg'
Learn More
Learn More
C16orf13 Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 20801-1-AP-20UL
This item is not returnable.
View return policy
Specifications
| C16orf13 | |
| Polyclonal | |
| Unconjugated | |
| METTL26 | |
| C16orf13, JFP2, UPF0585 protein C16orf13 | |
| Rabbit | |
| Antigen Affinity Chromatography | |
| RUO | |
| 302998, 68347, 84326 | |
| -20°C | |
| Liquid |
| Western Blot, Immunofluorescence, Immunocytochemistry | |
| 0.31 mg/mL | |
| PBS with 50% glycerol and 0.1% sodium azide; pH 7.3 | |
| Q497C3, Q96S19, Q9DCS2 | |
| Mettl26 | |
| C16orf13 Fusion Protein Ag14718 | |
| 20 μL | |
| Primary | |
| Mouse, Human, Rat | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction