missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C15orf52 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559747
This item is not returnable.
View return policy
Description
Immunogen sequence: ALLQPDGLTV TISQVPGEKR VVSRNWARGT CGPRVTNEML EDEDAEDHGG TFCLGELVEL AVTMENKAEG KRIVSEK Highest antigen sequence identity to the following orthologs: Mouse - 73%, Rat - 70%.
Specifications
| C15orf52 | |
| Polyclonal | |
| Unconjugated | |
| CCDC9B | |
| C15orf52; CCDC9B; chromosome 15 open reading frame 52; Coiled-coil domain-containing protein 9B; uncharacterized protein C15orf52 | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 388115 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry | |
| 0.4 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q6ZUT6 | |
| CCDC9B | |
| Recombinant protein corresponding to Human C15orf52. Recombinant protein control fragment (Product #RP-98184). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction