missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Invitrogen™ C15orf27 Polyclonal Antibody

Rabbit Polyclonal Antibody
Brand: Invitrogen™ PA559788
This item is not returnable.
View return policy
Description
Immunogen sequence: SLSEDPCPSQ KALDPAPLAR PSPAGSAQTS PELEHRVSLF NQKNQEGFTV FQIRPVIHFQ PTVPMLEDKF RSLESKEQKL HRV Highest antigen sequence identity to the following orthologs: Mouse - 78%, Rat - 78%.
C15orf27 is a voltage-sensor protein present on the post-synaptic side of glutamatergic mossy fibers and granule cells in the cerebellum. Despite the presence of a voltage-sensor segment, it does not form a functional ion channel and its precise role remains unclear. The protein undergoes both rapid and slow structural rearrangements in response to changes in voltage, and contains a zinc-binding site that can regulate the slow conformational transition.
Specifications
| C15orf27 | |
| Polyclonal | |
| Unconjugated | |
| TMEM266 | |
| 9630029F15; AI118078; C15orf27; expressed sequence AI118078; HsHVRP1; hTMEM266; Hv1 related protein 1; HV1-related protein 1; HVRP1; Tmem266; Transmembrane protein 266; transmembrane protein C15orf27; transmembrane protein C15orf27 homolog | |
| Rabbit | |
| Antigen affinity chromatography | |
| RUO | |
| 123591 | |
| Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles. | |
| Liquid |
| Immunohistochemistry (Paraffin), Immunocytochemistry | |
| 1 mg/mL | |
| PBS with 40% glycerol and 0.02% sodium azide; pH 7.2 | |
| Q2M3C6 | |
| TMEM266 | |
| Recombinant protein corresponding to Human C15orf27. Recombinant protein control fragment (Product #RP-98127). | |
| 100 μL | |
| Primary | |
| Human | |
| Antibody | |
| IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction