missing translation for 'onlineSavingsMsg'
Learn More

Invitrogen™ C14orf93 Monoclonal Antibody (2-F6)
GREENER_CHOICE

Product Code. 16326294
Change view
Click to view available options
Quantity:
100 μL
Unit Size:
100µL
1 product options available for selection
Product selection table with 1 available options. Use arrow keys to navigate and Enter or Space to select.
Product Code. Quantity unitSize
16326294 100 μL 100µL
Use arrow keys to navigate between rows. Press Enter or Space to select a product option. 1 options available.
1 options
This item is not returnable. View return policy
Product Code. 16326294 Supplier Invitrogen™ Supplier No. MA532882

Please to purchase this item. Need a web account? Register with us today!

This item is not returnable. View return policy

Mouse Monoclonal Antibody

C14orf93 Monoclonal Antibody for Western Blot, ICC/IF, IHC (P)

C14orf93 is a protein coding gene.
TRUSTED_SUSTAINABILITY

Specifications

Antigen C14orf93
Applications Immunohistochemistry (Paraffin), Western Blot, Immunocytochemistry, Western Blot
Classification Monoclonal
Clone 2-F6
Concentration 2 mg/mL
Conjugate Unconjugated
Formulation PBS with 0.2% BSA, 50% Glycerol and 0.05% sodium azide; pH 7.4
Gene C14orf93
Gene Accession No. Q9H972
Gene Alias C14orf93; chromosome 14 open reading frame 93; uncharacterized protein C14orf93
Gene Symbols C14orf93
Host Species Mouse
Immunogen Recombinant protein within Human C14orf93 aa 1-263.
Purification Method Protein A
Quantity 100 μL
Regulatory Status RUO
Primary or Secondary Primary
Gene ID (Entrez) 60686
Target Species Human
Content And Storage Store at 4°C short term. For long term storage, store at -20°C, avoiding freeze/thaw cycles.
Product Type Antibody
Form Liquid
Isotype IgG1
Show More Show Less
Product Title
Select an issue

By clicking Submit, you acknowledge that you may be contacted by Fisher Scientific in regards to the feedback you have provided in this form. We will not share your information for any other purposes. All contact information provided shall also be maintained in accordance with our Privacy Policy.